Assay Details
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004083.1 | NP_001004083.1 | 2 - 3 | 1136 | 74 | |
XM_006228575.2 | XP_006228637.1 | 2 - 3 | 1185 | 74 | ||
GenBank mRNA | AY229997.1 | - | 1 - 2 | 866 | 74 | |
AY952204.1 | - | 1 - 2 | 866 | 74 | ||
BC078907.1 | - | 2 - 3 | 1136 | 74 | ||
BC081858.1 | - | 2 - 3 | 1159 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
