Assay Details
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004083.1 | NP_001004083.1 | 4 - 5 | 1390 | 112 | |
XM_006228574.2 | XP_006228636.1 | 4 - 5 | 1400 | 112 | ||
XM_006228575.2 | XP_006228637.1 | 4 - 5 | 1439 | 112 | ||
GenBank mRNA | AY952204.1 | - | 3 - 4 | 1120 | 112 | |
BC078907.1 | - | 4 - 5 | 1390 | 112 | ||
BC081858.1 | - | 4 - 5 | 1413 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
