Assay Details
Gene Symbol: Grin2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022797.1 | NP_073634.1 | 8 - 9 | 2596 | 141 | |
XM_008759316.1 | XP_008757538.1 | 7 - 8 | 2301 | 141 | ||
XM_008759317.1 | XP_008757539.1 | 7 - 8 | 2301 | 141 | ||
GenBank mRNA | D13213.1 | - | 7 - 8 | 1890 | 141 | |
L31611.1 | - | 7 - 8 | 1871 | 141 | ||
L31612.1 | - | 8 - 9 | 2596 | 141 | ||
U08260.1 | - | 8 - 9 | 1942 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
