Assay Details
Gene Symbol: Dyrk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107496.1 | NP_001100966.1 | 7 - 8 | 1209 | 60 | |
XM_006228621.2 | XP_006228683.1 | 7 - 8 | 1373 | 60 | ||
XM_006228622.2 | XP_006228684.1 | 7 - 8 | 1373 | 60 | ||
XM_006228623.2 | XP_006228685.1 | 7 - 8 | 1373 | 60 | ||
XM_006228624.2 | XP_006228686.1 | 7 - 8 | 1224 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
