Assay Details
Gene Symbol: Tph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100634.2 | NP_001094104.1 | 6 - 7 | 582 | 125 | |
XM_006229218.2 | XP_006229280.1 | 5 - 6 | 708 | 125 | ||
XM_006229219.1 | XP_006229281.1 | 6 - 7 | 621 | 125 | ||
GenBank mRNA | CB809951.1 | - | 2 - 3 | 145 | 125 | |
X53501.1 | - | 6 - 7 | 582 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
