Assay Details
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031139.1 | NP_112401.1 | 7 - 8 | 1238 | 72 | |
GenBank mRNA | AB035647.1 | - | 7 - 8 | 727 | 72 | |
AB035648.1 | - | 7 - 8 | 646 | 72 | ||
AB035649.1 | - | 6 - 7 | 526 | 72 | ||
AB035650.1 | - | 6 - 7 | 680 | 72 | ||
AB035651.1 | - | 4 - 5 | 360 | 72 | ||
AB047556.1 | - | 7 - 8 | 1238 | 72 | ||
BC128735.1 | - | 7 - 8 | 732 | 7190049 | 72 | |
X90823.1 | - | 5 - 6 | 562 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
