Assay Details
Gene Symbol: Ppp1r13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228495.2 | XP_006228557.1 | 9 - 10 | 4829 | 94 | |
XM_008759052.1 | XP_008757274.1 | 10 - 11 | 4364 | 94 | ||
XM_008759053.1 | XP_008757275.1 | 9 - 10 | 3533 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
