Assay Details
Gene Symbol: Lsm14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127552.1 | NP_001121024.1 | 2 - 3 | 389 | 76 | |
XM_006228856.2 | XP_006228918.1 | 2 - 3 | 437 | 76 | ||
XM_006228857.2 | XP_006228919.1 | 2 - 3 | 437 | 76 | ||
XM_006228858.2 | XP_006228920.1 | 2 - 3 | 437 | 76 | ||
GenBank mRNA | BC166537.1 | - | 2 - 3 | 389 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
