Assay Details
Gene Symbol: Lsm14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127552.1 | NP_001121024.1 | 8 - 9 | 1246 | 103 | |
XM_006228856.2 | XP_006228918.1 | 8 - 9 | 1294 | 103 | ||
XM_006228857.2 | XP_006228919.1 | 8 - 9 | 1294 | 103 | ||
XM_006228858.2 | XP_006228920.1 | 7 - 8 | 1117 | 103 | ||
GenBank mRNA | BC166537.1 | - | 8 - 9 | 1246 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P002061 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1047615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64156 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1047615 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002061 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002061 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1047615 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1047615 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P002061 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1047615 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P478002 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8914 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10721036 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721031 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721030 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721033 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721037 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721034 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721035 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721029 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721032 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10721028 | - | - |
Rat Genome 230 2.0 Array | 1377905_at | - | - |
Rat Genome 230 2.0 Array | 1377905_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381970_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381970_at | - | - |
Affymetrix HT Rat Focus Array | 1377905_at | - | - |
Affymetrix HT Rat Focus Array | 1381970_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377905_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1381970_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100042105.rn.1 | TC0100005201.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100042111.rn.1 | TC0100005201.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100042106.rn.1 | TC0100005201.rn.1 | RTA TC |
Rat Expression Set 230 | 1381970_at | - | - |
Rat Expression Set 230 | 1381970_PM_at | - | - |
Rat Expression Set 230 | 1377905_at | - | - |
Rat Expression Set 230 | 1377905_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10721028 | - | - |
Rat Genome U34 Set | rc_AA875559_at | - | - |
Rat Genome U34 Set | rc_AA859351_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21914554 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370830 | - | - |
