Assay Details
Gene Symbol: Klc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138520.1 | NP_612529.1 | 3 - 4 | 516 | 154 | |
XM_006228377.2 | XP_006228439.1 | 3 - 4 | 669 | 154 | ||
XM_006228378.2 | XP_006228440.1 | 3 - 4 | 669 | 154 | ||
XM_006228379.2 | XP_006228441.1 | 3 - 4 | 591 | 154 | ||
XM_006228380.2 | XP_006228442.1 | 3 - 4 | 591 | 154 | ||
GenBank mRNA | AF166267.1 | - | 3 - 4 | 516 | 154 | |
BC078736.1 | - | 3 - 4 | 529 | 154 | ||
BC097284.1 | - | 3 - 4 | 521 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
