Assay Details
Gene Symbol: Tead2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107512.1 | NP_001100982.1 | 5 - 6 | 591 | 62 | |
XM_006229069.1 | XP_006229131.1 | 5 - 6 | 546 | 62 | ||
XM_006229070.2 | XP_006229132.1 | 5 - 6 | 548 | 62 | ||
XM_008759378.1 | XP_008757600.1 | 5 - 6 | 630 | 62 | ||
GenBank mRNA | BC168947.1 | - | 5 - 6 | 540 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
