Assay Details
Gene Symbol: Eva1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_340966.7 | XP_340967.4 | 1 - 2 | 1274 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
