Assay Details
Gene Symbol: Mrpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768604.1 | XP_008766826.1 | 1 - 2 | 116 | 97 | |
GenBank mRNA | FQ225027.1 | - | 1 - 2 | 75 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768604.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
