Assay Details
Gene Symbol: Mrpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768604.1 | XP_008766826.1 | 4 - 5 | 747 | 130 | |
GenBank mRNA | FQ225027.1 | - | 4 - 5 | 706 | 130 | |
FQ230552.1 | - | 4 - 5 | 707 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
