Assay Details
Gene Symbol: Mrpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768604.1 | XP_008766826.1 | 6 - 7 | 967 | 69 | |
GenBank mRNA | FQ225027.1 | - | 6 - 7 | 926 | 69 | |
FQ230552.1 | - | 6 - 7 | 927 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
