Assay Details
Gene Symbol: Il10rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107111.1 | NP_001100581.1 | 4 - 5 | 541 | 97 | |
XM_006248062.2 | XP_006248124.1 | 4 - 5 | 770 | 97 | ||
XM_006248063.2 | XP_006248125.1 | 4 - 5 | 1114 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
