Assay Details
Gene Symbol: Il10rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107111.1 | NP_001100581.1 | 6 - 7 | 847 | 82 | |
XM_006248062.2 | XP_006248124.1 | 6 - 7 | 1079 | 82 | ||
XM_006248063.2 | XP_006248125.1 | 6 - 7 | 1423 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
