Assay Details
Gene Symbol: Tm6sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108490.1 | NP_001101960.1 | 6 - 7 | 617 | 82 | |
XM_006229441.2 | XP_006229503.1 | 5 - 6 | 499 | 82 | ||
XM_008759543.1 | XP_008757765.1 | 4 - 5 | 879 | 82 | ||
XM_008759544.1 | XP_008757766.1 | 5 - 6 | 471 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
