Assay Details
Gene Symbol: Tm6sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108490.1 | NP_001101960.1 | 3 - 4 | 328 | 100 | |
XM_006229441.2 | XP_006229503.1 | 2 - 3 | 210 | 100 | ||
XM_008759544.1 | XP_008757766.1 | 2 - 3 | 182 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008759544.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
