Assay Details
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053309.1 | NP_445761.1 | 7 - 8 | 1069 | 92 | |
GenBank mRNA | AB007689.1 | - | 7 - 8 | 1069 | 92 | |
AB007690.1 | - | 7 - 8 | 850 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
