Assay Details
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053309.1 | NP_445761.1 | 8 - 9 | 1145 | 113 | |
GenBank mRNA | AB007689.1 | - | 8 - 9 | 1145 | 113 | |
AB007690.1 | - | 8 - 9 | 926 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G protein-coupled glutamate receptor signaling pathway
regulation of G protein-coupled receptor signaling pathway
calcium-mediated signaling using intracellular calcium source
behavioral response to cocaine
chemical homeostasis within a tissue
regulation of store-operated calcium entry
actin binding
protein binding
protein domain specific binding
synaptic receptor adaptor activity
glutamate receptor binding
G protein-coupled glutamate receptor binding
protein homodimerization activity
protein heterodimerization activity
G protein-coupled glutamate receptor signaling pathway
regulation of G protein-coupled receptor signaling pathway
calcium-mediated signaling using intracellular calcium source
behavioral response to cocaine
chemical homeostasis within a tissue
regulation of store-operated calcium entry
actin binding
protein binding
protein domain specific binding
synaptic receptor adaptor activity
glutamate receptor binding
G protein-coupled glutamate receptor binding
protein homodimerization activity
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P016059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140984 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P016059 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P016059 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P016059 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P640641 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702199977 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64068 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66296 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59347 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27068 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21854 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89077 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15522 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702199977 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10723309 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723305 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723301 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723313 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723308 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723314 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723302 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723303 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723311 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723310 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723300 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723304 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723312 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723306 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723307 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10723299 | - | - |
Rat Genome 230 2.0 Array | 1368872_a_at | - | - |
Rat Genome 230 2.0 Array | 1368872_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368872_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368872_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100047183.rn.1 | TC0100005908.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100047182.rn.1 | TC0100005908.rn.1 | RTA TC |
Rat Expression Set 230 | 1368872_a_at | - | - |
Rat Expression Set 230 | 1368872_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10723299 | - | - |
Rat Genome U34 Set | AB007689_at | - | - |
Rat Genome U34 Set | AB007690_s_at | - | - |
Rat Genome U34 Set | AB007689_g_at | - | - |
Rat Genome U34 Set | rc_AI058957_s_at | - | - |
Rat Genome U34 Set | rc_AA817674_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21140684 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365351 | - | - |
