Assay Details
Gene Symbol: Asb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108915.1 | NP_001102385.1 | 1 - 2 | 452 | 79 | |
XM_006229349.2 | XP_006229411.1 | 1 - 2 | 530 | 79 | ||
XM_006229350.2 | XP_006229412.1 | 1 - 2 | 530 | 79 | ||
XM_008759545.1 | XP_008757767.1 | 1 - 2 | 530 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108915.1, XM_006229349.2, XM_006229350.2, XM_008759545.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
