Assay Details
Gene Symbol: Asb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108915.1 | NP_001102385.1 | 2 - 3 | 537 | 116 | |
XM_006229349.2 | XP_006229411.1 | 2 - 3 | 615 | 116 | ||
XM_008759545.1 | XP_008757767.1 | 2 - 3 | 615 | 116 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001108915.1, XM_006229349.2, XM_008759545.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
