Assay Details
Gene Symbol: Fah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017181.2 | NP_058877.1 | 9 - 10 | 941 | 75 | |
GenBank mRNA | BC076381.1 | - | 9 - 10 | 941 | 75 | |
FQ219602.1 | - | 9 - 10 | 900 | 76 | ||
M77694.1 | - | 9 - 10 | 863 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
