Assay Details
Gene Symbol: Sh3gl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031238.2 | NP_112517.1 | 1 - 2 | 390 | 68 | |
GenBank mRNA | AF227439.1 | - | 1 - 2 | 393 | 68 | |
BC078800.1 | - | 1 - 2 | 393 | 7135194 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF227439.1, BC078800.1, NM_031238.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
