Assay Details
Gene Symbol: Sh3gl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031238.2 | NP_112517.1 | 6 - 7 | 971 | 130 | |
XM_006229480.2 | XP_006229542.1 | 6 - 7 | 759 | 130 | ||
XM_006229481.2 | XP_006229543.1 | 6 - 7 | 852 | 130 | ||
XM_006229483.2 | XP_006229545.1 | 6 - 7 | 958 | 130 | ||
XM_006229485.1 | XP_006229547.1 | 4 - 5 | 502 | 130 | ||
XM_006229486.2 | XP_006229548.1 | 4 - 5 | 426 | 130 | ||
GenBank mRNA | AB008159.2 | - | 3 - 4 | 416 | 130 | |
AF009604.1 | - | 4 - 5 | 455 | 130 | ||
AF227439.1 | - | 6 - 7 | 974 | 130 | ||
BC078800.1 | - | 6 - 7 | 974 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
