Assay Details
Gene Symbol: Cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031145.1 | NP_112407.1 | 4 - 5 | 383 | 78 | |
GenBank mRNA | AF136585.1 | - | 4 - 5 | 383 | 78 | |
BC091143.1 | - | 4 - 5 | 369 | 7308828 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
