Assay Details
Gene Symbol: Zfand6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229504.1 | XP_006229566.1 | 2 - 3 | 232 | 65 | |
XM_008759611.1 | XP_008757833.1 | 1 - 2 | 1623 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006229504.1, XM_008759611.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
