Assay Details
Gene Symbol: Mfge8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040186.2 | NP_001035276.1 | 2 - 3 | 272 | 106 | |
NM_012811.3 | NP_036943.1 | 2 - 3 | 272 | 106 | ||
GenBank mRNA | BC085754.1 | - | 2 - 3 | 225 | 106 | |
CO404762.1 | - | 2 - 3 | 274 | 106 | ||
D84068.1 | - | 2 - 3 | 232 | 106 | ||
DQ455823.1 | - | 2 - 3 | 225 | 106 | ||
FQ134652.1 | - | 2 - 3 | 272 | 106 | ||
FQ151276.1 | - | 2 - 3 | 272 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
