Assay Details
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012832.3 | NP_036964.3 | 8 - 9 | 879 | 76 | |
XM_008759497.1 | XP_008757719.1 | 6 - 7 | 4737 | 76 | ||
XM_008759498.1 | XP_008757720.1 | 9 - 10 | 1026 | 76 | ||
GenBank mRNA | AY574256.1 | - | 8 - 9 | 879 | 76 | |
AY671974.1 | - | 9 - 10 | 969 | 76 | ||
L31619.1 | - | 8 - 9 | 901 | 76 | ||
S53987.1 | - | 920 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
positive regulation of heart rate involved in baroreceptor response to decreased systemic arterial blood pressure
ion transport
calcium ion transport
cellular calcium ion homeostasis
endocytosis
signal transduction
synaptic transmission, cholinergic
memory
positive regulation of cell population proliferation
associative learning
regulation of norepinephrine secretion
neuronal action potential
flagellated sperm motility
response to food
regulation of synaptic transmission, dopaminergic
negative regulation of interleukin-1 beta production
negative regulation of interleukin-6 production
negative regulation of tumor necrosis factor production
ion transmembrane transport
response to nicotine
behavioral response to nicotine
T cell activation
B cell activation
regulation of membrane potential
dopamine biosynthetic process
ovulation cycle
response to ethanol
positive regulation of angiogenesis
behavioral response to ethanol
regulation of inflammatory response
negative regulation of inflammatory response
cognition
generation of ovulation cycle rhythm
cation transmembrane transport
amyloid-beta binding
protein binding
adenylate cyclase binding
acetylcholine receptor activity
toxic substance binding
chloride channel regulator activity
protein kinase binding
acetylcholine-gated cation-selective channel activity
acetylcholine binding
protein homodimerization activity
ATPase binding
scaffold protein binding
response to hypoxia
positive regulation of heart rate involved in baroreceptor response to decreased systemic arterial blood pressure
ion transport
calcium ion transport
cellular calcium ion homeostasis
endocytosis
signal transduction
synaptic transmission, cholinergic
memory
positive regulation of cell population proliferation
associative learning
regulation of norepinephrine secretion
neuronal action potential
flagellated sperm motility
response to food
regulation of synaptic transmission, dopaminergic
negative regulation of interleukin-1 beta production
negative regulation of interleukin-6 production
negative regulation of tumor necrosis factor production
ion transmembrane transport
response to nicotine
behavioral response to nicotine
T cell activation
B cell activation
regulation of membrane potential
dopamine biosynthetic process
ovulation cycle
response to ethanol
positive regulation of angiogenesis
behavioral response to ethanol
regulation of inflammatory response
negative regulation of inflammatory response
cognition
generation of ovulation cycle rhythm
cation transmembrane transport
amyloid-beta binding
protein binding
adenylate cyclase binding
acetylcholine receptor activity
toxic substance binding
chloride channel regulator activity
protein kinase binding
acetylcholine-gated cation-selective channel activity
acetylcholine binding
protein homodimerization activity
ATPase binding
scaffold protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P085087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65894 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085087 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085087 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P085087 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P288142 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11579 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16274 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97241 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90030 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95396 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88926 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58985 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45335 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13100 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69894 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47378 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4774 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42552 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15867 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17752 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10722543 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722545 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722536 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722542 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722533 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722537 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722538 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722534 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722544 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722539 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722535 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10722532 | - | - |
Rat Genome 230 2.0 Array | 1395429_at | - | - |
Rat Genome 230 2.0 Array | 1387419_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387419_at | - | - |
Rat Genome 230 2.0 Array | 1395429_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387419_at | - | - |
Affymetrix HT Rat Focus Array | 1395429_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387419_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395429_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100045509.rn.1 | TC0100005665.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100045510.rn.1 | TC0100005665.rn.1 | RTA TC |
Rat Expression Set 230 | 1387419_PM_at | - | - |
Rat Expression Set 230 | 1387419_at | - | - |
Rat Expression Set 230 | 1395429_PM_at | - | - |
Rat Expression Set 230 | 1395429_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722532 | - | - |
Rat Genome U34 Set | L31619_g_at | - | - |
Rat Genome U34 Set | L31619_at | - | - |
Rat Genome U34 Set | S53987_at | - | - |
Rat Genome U34 Set | L31619_at | - | - |
Rat Genome U34 Set | S53987_at | - | - |
Rat Genome U34 Set | L31619_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21334418 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040776 | - | - |
RatRef-12 Expression BeadChip | ILMN_66094 | - | - |
