Assay Details
Gene Symbol: Mrps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109148.1 | NP_001102618.1 | 4 - 5 | 474 | 58 | |
XM_006229400.1 | XP_006229462.1 | 5 - 6 | 429 | 58 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
