Assay Details
Gene Symbol: Dusp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022199.1 | NP_071535.1 | 2 - 3 | 577 | 103 | |
GenBank mRNA | U23438.1 | - | 2 - 3 | 577 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
