Assay Details
Gene Symbol: Cabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_574595.6 | XP_574595.3 | 3 - 4 | 945 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
