Assay Details
Gene Symbol: Lpxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009649.1 | NP_001009649.1 | 5 - 6 | 562 | 100 | |
GenBank mRNA | BC088217.1 | - | 5 - 6 | 562 | 7384057 | 100 |
FQ230386.1 | - | 5 - 6 | 527 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
