Assay Details
Gene Symbol: Aldh1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022407.3 | NP_071852.2 | 3 - 4 | 337 | 72 | |
GenBank mRNA | AF001896.1 | - | 3 - 4 | 337 | 72 | |
AF001897.1 | - | 3 - 4 | 337 | 72 | ||
AF001898.1 | - | 3 - 4 | 337 | 72 | ||
BC061526.1 | - | 3 - 4 | 356 | 6921926 | 72 | |
U79118.1 | - | 3 - 4 | 336 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
