Assay Details
Gene Symbol: Mtg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749002.3 | XP_003749050.1 | 9 - 10 | 803 | 85 | |
XM_006230584.2 | XP_006230646.1 | 9 - 10 | 819 | 85 |
Gene Symbol: Paox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF329824.1 | - | 86 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
