Assay Details
Gene Symbol: Tm7sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013071.1 | NP_001013089.1 | 1 - 2 | 139 | 108 | |
GenBank mRNA | BC091237.1 | - | 1 - 2 | 139 | 108 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC091237.1, NM_001013071.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
