Assay Details
Gene Symbol: Sipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004089.1 | NP_001004089.1 | 2 - 3 | 802 | 77 | |
XM_006230849.2 | XP_006230911.1 | 3 - 4 | 1419 | 77 | ||
GenBank mRNA | AB091474.1 | - | 2 - 3 | 802 | 77 | |
FQ223350.1 | - | 3 - 4 | 1009 | 77 | ||
FQ228273.1 | - | 3 - 4 | 988 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
