Assay Details
Gene Symbol: Slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013032.3 | NP_037164.3 | 4 - 5 | 528 | 74 | |
GenBank mRNA | BC061743.1 | - | 4 - 5 | 528 | 74 | |
D63772.1 | - | 559 | 74 | |||
U21104.1 | - | 4 - 5 | 540 | 74 | ||
X94255.1 | - | 4 - 5 | 585 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
