Assay Details
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005907.2 | NP_001005907.1 | 8 - 9 | 959 | 82 | |
NM_001277341.1 | NP_001264270.1 | 9 - 10 | 1082 | 82 | ||
NM_001277346.1 | NP_001264275.1 | 7 - 8 | 752 | 82 | ||
XM_006230742.2 | XP_006230804.1 | 9 - 10 | 1422 | 82 | ||
XM_006230743.1 | XP_006230805.1 | 9 - 10 | 1073 | 82 | ||
XM_006230744.2 | XP_006230806.1 | 9 - 10 | 1079 | 82 | ||
GenBank mRNA | BC083804.1 | - | 8 - 9 | 897 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
