Assay Details
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005907.2 | NP_001005907.1 | 2 - 3 | 216 | 63 | |
NM_001277341.1 | NP_001264270.1 | 2 - 3 | 216 | 63 | ||
NM_001277346.1 | NP_001264275.1 | 2 - 3 | 216 | 63 | ||
XM_006230742.2 | XP_006230804.1 | 2 - 3 | 556 | 63 | ||
XM_006230743.1 | XP_006230805.1 | 2 - 3 | 207 | 63 | ||
XM_006230744.2 | XP_006230806.1 | 2 - 3 | 213 | 63 | ||
GenBank mRNA | BC083804.1 | - | 2 - 3 | 154 | 63 | |
CB811907.1 | - | 2 - 3 | 216 | 63 | ||
FM037652.1 | - | 2 - 3 | 178 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
