Assay Details
Gene Symbol: Fuom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106310.1 | NP_001099780.1 | 2 - 3 | 592 | 58 | |
GenBank mRNA | FQ212771.1 | - | 2 - 3 | 174 | 58 |
Gene Symbol: LOC100911225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748887.3 | XP_003748935.1 | 2 - 3 | 214 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
