Assay Details
Gene Symbol: Lipf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017341.2 | NP_059037.1 | 4 - 5 | 464 | 100 | |
GenBank mRNA | A01157.1 | - | 3 - 4 | 423 | 100 | |
X02309.1 | - | 3 - 4 | 465 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
