Assay Details
Gene Symbol: Zfand5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760297.1 | XP_008758519.1 | 2 - 3 | 741 | 77 | |
GenBank mRNA | BC168882.1 | - | 2 - 3 | 209 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
