Assay Details
Gene Symbol: Tmem80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045202.1 | 4 - 5 | 973 | 142 | ||
GenBank mRNA | CB546081.1 | - | 4 - 5 | 320 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
