Assay Details
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012861.1 | NP_036993.1 | 1 - 2 | 64 | 80 | |
GenBank mRNA | S61804.1 | - | 1 - 2 | 64 | 80 | |
X54862.1 | - | 1 - 2 | 90 | 80 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_012861.1, S61804.1, X54862.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
