Assay Details
Gene Symbol: Lgals12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106333.1 | NP_001099803.1 | 2 - 3 | 620 | 120 | |
XM_006230763.2 | XP_006230825.1 | 3 - 4 | 1187 | 120 | ||
XM_006230764.2 | XP_006230826.1 | 3 - 4 | 450 | 120 | ||
XM_008760110.1 | XP_008758332.1 | 3 - 4 | 1399 | 120 | ||
XM_008760111.1 | XP_008758333.1 | 3 - 4 | 1383 | 120 | ||
XM_008760112.1 | XP_008758334.1 | 2 - 3 | 549 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
