Assay Details
Gene Symbol: Lgals12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106333.1 | NP_001099803.1 | 8 - 9 | 1258 | 158 | |
XM_006230763.2 | XP_006230825.1 | 9 - 10 | 1825 | 158 | ||
XM_006230764.2 | XP_006230826.1 | 9 - 10 | 1088 | 158 | ||
XM_008760110.1 | XP_008758332.1 | 9 - 10 | 2037 | 158 | ||
XM_008760111.1 | XP_008758333.1 | 9 - 10 | 2021 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
