Assay Details
Gene Symbol: Stambpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115048.1 | NP_001108520.1 | 9 - 10 | 1443 | 63 | |
XM_006231296.2 | XP_006231358.1 | 9 - 10 | 1490 | 63 | ||
XM_006231297.2 | XP_006231359.1 | 9 - 10 | 1255 | 63 | ||
XM_006231299.2 | XP_006231361.1 | 9 - 10 | 1627 | 63 | ||
XM_008760346.1 | XP_008758568.1 | 8 - 9 | 1212 | 63 | ||
XM_008760347.1 | XP_008758569.1 | 7 - 8 | 1468 | 63 | ||
XM_008760349.1 | XP_008758571.1 | 8 - 9 | 1243 | 63 | ||
GenBank mRNA | BC158624.1 | - | 9 - 10 | 1443 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
