Assay Details
Gene Symbol: Glis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760357.1 | XP_008758579.1 | 7 - 8 | 929 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
